Assay Details
| Gene Symbol: LOC4336762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778933.2 | XP_015634419.1 | 4 - 5 | 565 | 81 | |
| GenBank mRNA | AK070540.1 | - | 4 - 5 | 464 | 81 | |
| AY551918.1 | - | 4 - 5 | 570 | 81 | ||
| FJ668596.1 | - | 4 - 5 | 498 | 81 | ||
More Information
Set Membership: |
|

LOC4336762
Genomic Map