Assay Details
| Gene Symbol: LOC4336762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778933.2 | XP_015634419.1 | 5 - 6 | 600 | 64 | |
| GenBank mRNA | AK070540.1 | - | 5 - 6 | 499 | 64 | |
| AY551918.1 | - | 5 - 6 | 605 | 64 | ||
| FJ668596.1 | - | 5 - 6 | 533 | 64 | ||
More Information
Set Membership: |
|

LOC4336762
Genomic Map