Assay Details
| Gene Symbol: LOC4336921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781794.2 | XP_015637280.1 | 3 - 4 | 360 | 118 | |
| GenBank mRNA | AK060169.1 | - | 3 - 4 | 308 | 118 | |
| AK122125.1 | - | 3 - 4 | 321 | 118 | ||
More Information
Set Membership: |
|

LOC4336921
Genomic Map