Assay Details
Gene Symbol: LOC4336930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781745.2 | 5 - 6 | 988 | 115 | ||
GenBank mRNA | AK065931.1 | - | 5 - 6 | 675 | 115 | |
AK099996.1 | - | 5 - 6 | 596 | 115 | ||
AK121814.1 | - | 5 - 6 | 997 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
