Assay Details
| Gene Symbol: LOC4337014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779487.2 | XP_015634973.1 | 5 - 6 | 604 | 76 | |
| GenBank mRNA | AK103625.1 | - | 5 - 6 | 611 | 76 | |
More Information
Set Membership: |
|

LOC4337014
Genomic Map