Assay Details
| Gene Symbol: LOC4337022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777911.2 | XP_015633397.1 | 1 - 2 | 172 | 86 | |
| GenBank mRNA | AK066247.1 | - | 1 - 2 | 143 | 86 | |
More Information
Set Membership: |
|

LOC4337022
Genomic Map