Assay Details
| Gene Symbol: LOC4338673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782466.2 | XP_015637952.1 | 3 - 4 | 1635 | 121 | |
| GenBank mRNA | AK064352.1 | - | 2 - 3 | 682 | 121 | |
| AK101529.1 | - | 3 - 4 | 1515 | 121 | ||
More Information
Set Membership: |
|

LOC4338673
Genomic Map