Assay Details
| Gene Symbol: LOC4338673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782466.2 | XP_015637952.1 | 5 - 6 | 1890 | 102 | |
| GenBank mRNA | AK064352.1 | - | 4 - 5 | 937 | 102 | |
| AK101529.1 | - | 5 - 6 | 1770 | 102 | ||
More Information
Set Membership: |
|

LOC4338673
Genomic Map