Assay Details
| Gene Symbol: LOC4338710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784981.2 | XP_015640467.2 | 10 - 11 | 1940 | 99 | |
| GenBank mRNA | AK102894.1 | - | 10 - 11 | 1694 | 99 | |
More Information
Set Membership: |
|

LOC4338710
Genomic Map