Assay Details
| Gene Symbol: LOC4338712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785078.2 | XP_015640564.1 | 13 - 14 | 1709 | 77 | |
| GenBank mRNA | AK241913.1 | - | 1 - 2 | 132 | 77 | |
More Information
Set Membership: |
|

LOC4338712
Genomic Map