Assay Details
| Gene Symbol: LOC4338712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785078.2 | XP_015640564.1 | 14 - 15 | 1768 | 99 | |
| GenBank mRNA | AK241913.1 | - | 2 - 3 | 191 | 99 | |
More Information
Set Membership: |
|

LOC4338712
Genomic Map