Assay Details
| Gene Symbol: LOC4338712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785078.2 | XP_015640564.1 | 6 - 7 | 1000 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4338712| Gene Symbol: LOC4338712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785078.2 | XP_015640564.1 | 6 - 7 | 1000 | 72 | |
Set Membership: |
|