Assay Details
| Gene Symbol: LOC4339756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784229.1 | XP_015639715.1 | 4 - 5 | 1625 | 74 | |
| GenBank mRNA | AK071721.1 | - | 4 - 5 | 1608 | 74 | |
More Information
Set Membership: |
|

LOC4339756
Genomic Map