Assay Details
| Gene Symbol: LOC4339799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785360.2 | XP_015640846.1 | 1 | 1775 | 92 | |
| GenBank mRNA | AK064253.1 | - | 1 | 1449 | 92 | |
More Information
Set Membership: |
|

LOC4339799
Genomic Map