Assay Details
| Gene Symbol: LOC4340941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788427.1 | XP_015643913.1 | 2 - 3 | 485 | 102 | |
| GenBank mRNA | AK073531.1 | - | 2 - 3 | 468 | 102 | |
| AK099015.1 | - | 2 - 3 | 470 | 102 | ||
| AK099141.1 | - | 2 - 3 | 468 | 102 | ||
| AK099616.1 | - | 2 - 3 | 470 | 102 | ||
| AK100193.1 | - | 3 - 4 | 347 | 102 | ||
| AK104270.1 | - | 2 - 3 | 469 | 102 | ||
| AK104377.1 | - | 2 - 3 | 469 | 102 | ||
| AK104464.1 | - | 2 - 3 | 467 | 102 | ||
More Information
Set Membership: |
|

LOC4340941
Genomic Map