Assay Details
| Gene Symbol: LOC4337240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780204.2 | XP_015635690.1 | 4 - 5 | 698 | 80 | |
| GenBank mRNA | AK059441.1 | - | 4 - 5 | 498 | 80 | |
| AK069326.1 | - | 4 - 5 | 501 | 80 | ||
| BT129135.1 | - | 4 - 5 | 443 | 80 | ||
| BT131105.1 | - | 4 - 5 | 421 | 80 | ||
More Information
Set Membership: |
|

LOC4337240
Genomic Map