Assay Details
Gene Symbol: LOC4337265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781248.2 | XP_015636734.1 | 1 - 2 | 573 | 104 | |
XM_015781249.2 | XP_015636735.1 | 1 - 2 | 535 | 104 | ||
GenBank mRNA | AK068928.1 | - | 1 - 2 | 451 | 104 | |
AK071255.1 | - | 1 - 2 | 381 | 104 | ||
HQ324813.1 | - | 1 - 2 | 182 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
