Assay Details
| Gene Symbol: LOC4337442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777980.2 | XP_015633466.1 | 1 - 2 | 1041 | 66 | |
| XM_015777981.2 | XP_015633467.1 | 1 - 2 | 1042 | 66 | ||
| XM_015777982.2 | XP_015633468.1 | 1 - 2 | 1043 | 66 | ||
| GenBank mRNA | AK065300.1 | - | 1 - 2 | 1047 | 66 | |
More Information
Set Membership: |
|

LOC4337442
Genomic Map