Assay Details
| Gene Symbol: LOC4337442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777980.2 | XP_015633466.1 | 4 - 5 | 1500 | 102 | |
| XM_015777981.2 | XP_015633467.1 | 4 - 5 | 1501 | 102 | ||
| XM_015777982.2 | XP_015633468.1 | 4 - 5 | 1502 | 102 | ||
| GenBank mRNA | AK065300.1 | - | 4 - 5 | 1506 | 102 | |
More Information
Set Membership: |
|

LOC4337442
Genomic Map