Assay Details
| Gene Symbol: LOC4337502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780149.2 | XP_015635635.1 | 1 - 2 | 331 | 65 | |
| GenBank mRNA | AB125306.1 | - | 1 - 2 | 255 | 65 | |
| AK100269.1 | - | 1 - 2 | 287 | 65 | ||
| JF733763.1 | - | 1 - 2 | 122 | 65 | ||
| JN848807.1 | - | 1 - 2 | 122 | 65 | ||
More Information
Set Membership: |
|

LOC4337502
Genomic Map