Assay Details
| Gene Symbol: LOC4337502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780149.2 | XP_015635635.1 | 2 - 3 | 850 | 82 | |
| GenBank mRNA | AB125306.1 | - | 2 - 3 | 774 | 82 | |
| AK100269.1 | - | 2 - 3 | 806 | 82 | ||
| JF733763.1 | - | 2 - 3 | 641 | 82 | ||
| JN848807.1 | - | 2 - 3 | 641 | 82 | ||
More Information
Set Membership: |
|

LOC4337502
Genomic Map