Assay Details
| Gene Symbol: LOC4337557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783678.2 | XP_015639164.1 | 1 - 2 | 334 | 76 | |
| GenBank mRNA | AK069395.1 | - | 1 - 2 | 330 | 76 | |
More Information
Set Membership: |
|

LOC4337557
Genomic Map