Assay Details
| Gene Symbol: LOC4337557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783678.2 | XP_015639164.1 | 4 - 5 | 708 | 96 | |
| GenBank mRNA | AK069395.1 | - | 4 - 5 | 732 | 96 | |
More Information
Set Membership: |
|

LOC4337557
Genomic Map