Assay Details
| Gene Symbol: LOC4337557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783678.2 | XP_015639164.1 | 9 - 10 | 1184 | 101 | |
| GenBank mRNA | AK069395.1 | - | 7 - 8 | 970 | 101 | |
More Information
Set Membership: |
|

LOC4337557
Genomic Map