Assay Details
| Gene Symbol: LOC4337589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784990.2 | XP_015640476.1 | 3 - 4 | 384 | 59 | |
| GenBank mRNA | AK060179.1 | - | 3 - 4 | 390 | 59 | |
| AK066175.1 | - | 3 - 4 | 340 | 59 | ||
More Information
Set Membership: |
|

LOC4337589
Genomic Map