Assay Details
| Gene Symbol: LOC4337589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784990.2 | XP_015640476.1 | 6 - 7 | 989 | 149 | |
| GenBank mRNA | AK060179.1 | - | 6 - 7 | 995 | 149 | |
| AK066175.1 | - | 6 - 7 | 945 | 149 | ||
More Information
Set Membership: |
|

LOC4337589
Genomic Map