Assay Details
| Gene Symbol: LOC4337608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785076.1 | XP_015640562.1 | 1 - 2 | 245 | 85 | |
| GenBank mRNA | AK069313.1 | - | 1 - 2 | 238 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015785076.1 transcripts.
Set Membership: |
|

LOC4337608
Genomic Map