Assay Details
| Gene Symbol: LOC4337608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785076.1 | XP_015640562.1 | 2 - 3 | 382 | 77 | |
| GenBank mRNA | AK069313.1 | - | 2 - 3 | 375 | 77 | |
More Information
Set Membership: |
|

LOC4337608
Genomic Map