Assay Details
| Gene Symbol: LOC4337608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785076.1 | XP_015640562.1 | 4 - 5 | 480 | 109 | |
| GenBank mRNA | AK069313.1 | - | 4 - 5 | 473 | 109 | |
More Information
Set Membership: |
|

LOC4337608
Genomic Map