Assay Details
| Gene Symbol: LOC4337722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784055.2 | XP_015639541.1 | 8 - 9 | 1072 | 68 | |
| GenBank mRNA | AK058276.1 | - | 8 - 9 | 1036 | 68 | |
| AK064884.1 | - | 8 - 9 | 1038 | 68 | ||
| AK121623.1 | - | 8 - 9 | 1035 | 68 | ||
| DQ060684.1 | - | 8 - 9 | 1034 | 68 | ||
More Information
Set Membership: |
|

LOC4337722
Genomic Map