Assay Details
| Gene Symbol: LOC4337828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785195.2 | XP_015640681.1 | 13 - 14 | 2351 | 123 | |
| GenBank mRNA | AK072977.1 | - | 11 - 12 | 2351 | 123 | |
| AY180331.1 | - | 12 - 13 | 2102 | 123 | ||
More Information
Set Membership: |
|

LOC4337828
Genomic Map