Assay Details
| Gene Symbol: LOC4337828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785195.2 | XP_015640681.1 | 2 - 3 | 860 | 77 | |
| GenBank mRNA | AK063338.1 | - | 1 - 2 | 944 | 77 | |
| AK072977.1 | - | 1 - 2 | 944 | 77 | ||
| AY180331.1 | - | 1 - 2 | 611 | 77 | ||
More Information
Set Membership: |
|

LOC4337828
Genomic Map