Assay Details
| Gene Symbol: LOC4337828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785195.2 | XP_015640681.1 | 5 - 6 | 1376 | 146 | |
| GenBank mRNA | AK063338.1 | - | 4 - 5 | 1460 | 146 | |
| AK072977.1 | - | 4 - 5 | 1460 | 146 | ||
| AY180331.1 | - | 4 - 5 | 1127 | 146 | ||
More Information
Set Membership: |
|

LOC4337828
Genomic Map