Assay Details
| Gene Symbol: LOC4337828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785195.2 | XP_015640681.1 | 7 - 8 | 1881 | 94 | |
| GenBank mRNA | AK072977.1 | - | 6 - 7 | 1965 | 94 | |
| AY180331.1 | - | 6 - 7 | 1632 | 94 | ||
More Information
Set Membership: |
|

LOC4337828
Genomic Map