Assay Details
| Gene Symbol: LOC4337876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783994.1 | XP_015639480.1 | 26 - 27 | 3203 | 69 | |
| XM_015783995.1 | XP_015639481.1 | 26 - 27 | 3200 | 69 | ||
| XM_015783996.1 | XP_015639482.1 | 26 - 27 | 3197 | 69 | ||
| GenBank mRNA | AK065411.1 | - | 26 - 27 | 3185 | 69 | |
| BT132604.1 | - | 1 - 2 | 63 | 69 | ||
More Information
Set Membership: |
|

LOC4337876
Genomic Map