Assay Details
| Gene Symbol: LOC4338140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783963.2 | XP_015639449.1 | 12 - 13 | 3208 | 77 | |
| XM_015783965.2 | XP_015639451.1 | 11 - 12 | 2976 | 77 | ||
| GenBank mRNA | AK111685.1 | - | 1 - 2 | 96 | 77 | |
More Information
Set Membership: |
|

LOC4338140
Genomic Map