Assay Details
| Gene Symbol: LOC4338140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783963.2 | XP_015639449.1 | 17 - 18 | 3682 | 134 | |
| XM_015783965.2 | XP_015639451.1 | 16 - 17 | 3450 | 134 | ||
| GenBank mRNA | AK111685.1 | - | 6 - 7 | 570 | 134 | |
More Information
Set Membership: |
|

LOC4338140
Genomic Map