Assay Details
| Gene Symbol: LOC4338140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783963.2 | XP_015639449.1 | 19 - 20 | 3791 | 61 | |
| XM_015783965.2 | XP_015639451.1 | 18 - 19 | 3559 | 61 | ||
| GenBank mRNA | AK111685.1 | - | 8 - 9 | 679 | 61 | |
More Information
Set Membership: |
|

LOC4338140
Genomic Map