Assay Details
| Gene Symbol: LOC4338154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782831.2 | XP_015638317.1 | 2 - 3 | 551 | 154 | |
| GenBank mRNA | AK064229.1 | - | 2 - 3 | 550 | 154 | |
| AK101642.1 | - | 2 - 3 | 467 | 154 | ||
More Information
Set Membership: |
|

LOC4338154
Genomic Map