Assay Details
| Gene Symbol: LOC4338203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782530.2 | XP_015638016.1 | 4 - 5 | 1095 | 74 | |
| GenBank mRNA | AK073635.1 | - | 4 - 5 | 1017 | 74 | |
| AK104978.1 | - | 4 - 5 | 982 | 74 | ||
More Information
Set Membership: |
|

LOC4338203
Genomic Map