Assay Details
| Gene Symbol: LOC4338258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784954.1 | XP_015640440.1 | 1 - 2 | 226 | 123 | |
| GenBank mRNA | AK065530.1 | - | 1 - 2 | 152 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784954.1 transcripts.
Set Membership: |
|

LOC4338258
Genomic Map