Assay Details
| Gene Symbol: LOC4338258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784954.1 | XP_015640440.1 | 2 - 3 | 1361 | 132 | |
| XM_015784955.1 | XP_015640441.1 | 1 - 2 | 1076 | 132 | ||
| GenBank mRNA | AB110207.1 | - | 1 - 2 | 1076 | 132 | |
| AK065530.1 | - | 2 - 3 | 1287 | 132 | ||
More Information
Set Membership: |
|

LOC4338258
Genomic Map