Assay Details
| Gene Symbol: LOC4338324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781848.2 | XP_015637334.1 | 3 - 4 | 386 | 103 | |
| GenBank mRNA | AK070232.1 | - | 3 - 4 | 289 | 103 | |
| HQ013293.1 | - | 3 - 4 | 254 | 103 | ||
More Information
Set Membership: |
|

LOC4338324
Genomic Map