Assay Details
| Gene Symbol: LOC4338324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781848.2 | XP_015637334.1 | 5 - 6 | 529 | 90 | |
| GenBank mRNA | AK070232.1 | - | 5 - 6 | 432 | 90 | |
| HQ013293.1 | - | 5 - 6 | 397 | 90 | ||
More Information
Set Membership: |
|

LOC4338324
Genomic Map