Assay Details
| Gene Symbol: LOC4338324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781848.2 | XP_015637334.1 | 7 - 8 | 686 | 95 | |
| GenBank mRNA | AK070232.1 | - | 7 - 8 | 589 | 95 | |
| HQ013293.1 | - | 7 - 8 | 554 | 95 | ||
More Information
Set Membership: |
|

LOC4338324
Genomic Map