Assay Details
Gene Symbol: LOC4332232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK287988.1 | - | 1630 | 77 |
Gene Symbol: LOC4338385 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782020.2 | XP_015637506.1 | 2 - 3 | 234 | 77 | |
XM_015782021.2 | XP_015637507.1 | 2 - 3 | 342 | 77 | ||
GenBank mRNA | AK121220.1 | - | 2 - 3 | 215 | 77 | |
AK241054.1 | - | 2 - 3 | 214 | 77 | ||
BT130711.1 | - | 2 - 3 | 216 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
