Assay Details
| Gene Symbol: LOC4338842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785223.2 | XP_015640709.1 | 4 - 5 | 699 | 75 | |
| XM_015785224.2 | XP_015640710.1 | 4 - 5 | 703 | 75 | ||
| GenBank mRNA | AK100233.1 | - | 4 - 5 | 518 | 75 | |
More Information
Set Membership: |
|

LOC4338842
Genomic Map