Assay Details
| Gene Symbol: LOC4339082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782024.2 | XP_015637510.1 | 1 - 2 | 1594 | 95 | |
| GenBank mRNA | AB126087.1 | - | 1 - 2 | 606 | 95 | |
| AK070864.1 | - | 1 - 2 | 741 | 95 | ||
| AY281300.1 | - | 1 - 2 | 619 | 95 | ||
More Information
Set Membership: |
|

LOC4339082
Genomic Map