Assay Details
| Gene Symbol: LOC4339082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782024.2 | XP_015637510.1 | 2 - 3 | 1745 | 59 | |
| GenBank mRNA | AB126087.1 | - | 2 - 3 | 757 | 59 | |
| AK070864.1 | - | 2 - 3 | 892 | 59 | ||
| AY281300.1 | - | 2 - 3 | 770 | 59 | ||
More Information
Set Membership: |
|

LOC4339082
Genomic Map