Assay Details
| Gene Symbol: LOC4339133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784345.2 | XP_015639831.1 | 1 - 2 | 71 | 101 | |
| GenBank mRNA | AK065541.1 | - | 1 - 2 | 75 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784345.2 transcripts.
Set Membership: |
|

LOC4339133
Genomic Map